5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure
Thermo Scientific Chemicals

5-Bromo-4-chloro-3-indolyl-beta-D-galactoside, 99%, pure

CAS: 7240-90-6 | C14H15BrClNO6 | 408.63 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
Catalog number 227771000
Price (USD)
104.65
Special offer
Online exclusive
Ends: 28-Dec-2026
123.00
Save 18.35
Each
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
104.65
Special offer
Online exclusive
Ends: 28-Dec-2026
123.00
Save 18.35
Each
Chemical Identifiers
CAS7240-90-6
IUPAC NameX-gal
Molecular FormulaC23H41Cu2N5O5
InChI KeyGGILAJWFDJWIQO-UHFFFAOYSA-N
SMILES[Cu+].[Cu++].CC(O)O.CC(O)O.CN1CCN(CC2=CC(=CC(CN3CCN(C)CC3)=C2O)C#N)CC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=97.5 %
Specific optical rotation-60° to -72° (20°C, 589 nm) (c=1, DMF)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only