Digoxin, 95%
Digoxin, 95%
Digoxin, 95%
Thermo Scientific Chemicals

Digoxin, 95%

CAS: 20830-75-5 | C41H64O14 | 780.94 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 228120010
Price (EUR)
97,65
Online exclusive
108,00
Save 10,35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
97,65
Online exclusive
108,00
Save 10,35
Each
Chemical Identifiers
CAS20830-75-5
IUPAC Name4-[(1R,3aS,3bR,5aR,7S,9aS,9bS,11R,11aS)-7-{[(2R,4S,5S,6R)-5-{[(2S,4S,5S,6R)-5-{[(2S,4S,5S,6R)-4,5-dihydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-4-hydroxy-6-methyloxan-2-yl]oxy}-3a,11-dihydroxy-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-1-yl]-2,5-dihydrofuran-2-one
Molecular FormulaC41H64O14
InChI KeyLTMHDMANZUZIPE-PUGKRICDSA-N
SMILESC[C@H]1O[C@H](C[C@H](O)[C@@H]1O)O[C@H]1[C@@H](O)C[C@H](O[C@H]2[C@@H](O)C[C@H](O[C@H]3CC[C@@]4(C)[C@H](CC[C@@H]5[C@@H]4C[C@@H](O)[C@]4(C)[C@H](CC[C@]54O)C4=CC(=O)OC4)C3)O[C@@H]2C)O[C@@H]1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Specific optical rotationon dried substance
Infrared spectrumConforms
HPLC>=94 % (on dried substance)
Loss on drying=<1 % (500 mg) (vacuum)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only