alpha-Methyl-D-mannopyranoside, 99+%
alpha-Methyl-D-mannopyranoside, 99+%
alpha-Methyl-D-mannopyranoside, 99+%
Thermo Scientific Chemicals

alpha-Methyl-D-mannopyranoside, 99+%

alpha-Methyl-D-mannopyranoside, 99+%, Quantity: 500g, Packaging: Glass bottle, Gray or White, Melting Point: 186.0 deg.C to 194.0 deg.C, Molecular Weight: 194.18, Percent Purity: 99+%, Appearance: (1% aq. soln.) clear, colorless, Assay Percent Range: 99% min. (ELSD) (HPLC) | CAS: 617-04-9 | C7H14O6 | 194.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 229250250
Price (USD)
52.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.20
Save 9.55
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
52.65
Special offer
Online exclusive
Ends: 28-Dec-2026
62.20
Save 9.55
Each
Chemical Identifiers
CAS617-04-9
IUPAC Name2-(hydroxymethyl)-6-methoxyoxane-3,4,5-triol
Molecular FormulaC7H14O6
InChI KeyHOVAGTYPODGVJG-UHFFFAOYNA-N
SMILESCOC1OC(CO)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=99.0 %
Water=<0.5 %
Sulfated ash=<0.2 %
Specific optical rotation+77° to +81° (20°C, 589 nm) (c=10, water)
Solubility(10 % in water) Clear colorless
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only