Search
View additional product information for Pierce™ Phosphoprotein Phosphate Estimation Assay Kit - FAQs (23270)
2 product FAQs found
The Glycoprotein Carbohydrate Estimation Kit (Cat. No.23260) enables the amount of protein glycosylation to be measured as the percent of total purified protein mass.
The Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270) enables classification and identification of proteins as phosphorylated serine and threonine, as well as semi-quantitative assessment of the phosphorylation level.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.
We offer several specialty protein assays including: Protease assay kits (Cat. Nos. 23263 and 23266), Glycoprotein Carbohydrate Estimation Kit (Cat. No. 23260), Phosphoprotein Phosphate Estimation Kit (Cat. No. 23270), Quantitative Peroxide assay kits (Cat. Nos. 23280 and 23285), and Easy-Titer Assay kits for IgG and IgM.
Find additional tips, troubleshooting help, and resources within our Protein Assays and Analysis Support Center.