This product ships via ground from US northeast.
Polysorbate 20
Polysorbate 20
Polysorbate 20
Thermo Scientific Chemicals

Polysorbate 20

CAS: 9005-64-5 | C26H50O10
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
250 mL
1 L
5 L
25 L
Packaging Type:
View all
Glass Bottle
Catalog number 233362500
Price (USD)
52.60
Each
Quantity:
250 mL
Packaging Type:
Glass Bottle
Request bulk or custom format
Price (USD)
52.60
Each
Chemical Identifiers
CAS9005-64-5
Molecular Formula(C2H4O)y(C2H4O)w(C2H4O)x(C2H4O)zC18H34O6
InChI KeyHMFKFHLTUCJZJO-UHFFFAOYNA-N
SMILESCCCCCCCCCCCC(=O)OCCOCC(OCCO)C1OCC(OCCO)C1OCCO
MDL NumberMFCD00165986
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear to slightly hazy yellow to brown-yellow
Appearance (Form)Viscous liquid
AppearanceMay develop some turbidity or precipitate
Infrared spectrumConforms
Water=<3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only