Search
View additional product information for Pierce™ c-Myc-Tag IP/Co-IP Kit - FAQs (23620)
1 product FAQs found
We recommend using Pierce IP Lysis Buffer (Cat. No. 87787, 87788).
Find additional tips, troubleshooting help, and resources within our Protein Dialysis, Desalting, and Concentration Support Center.