Search
View additional product information for Nunc™ Square BioAssay Dishes - FAQs (166508, 240835, 240845)
1 product FAQs found
We offer Nunc Square BioAssay Dishes, but this product does not have a grid.