L(-)-Glucose, 98%
L(-)-Glucose, 98%
L(-)-Glucose, 98%
Thermo Scientific Chemicals

L(-)-Glucose, 98%

CAS: 921-60-8 | C6H12O6 | 180.16 g/mol
数量:
250 mg
1 g
5 g
25 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 241920250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS921-60-8
IUPAC Name6-(hydroxymethyl)oxane-2,3,4,5-tetrol
Molecular FormulaC6H12O6
InChI KeyWQZGKKKJIJFFOK-UHFFFAOYNA-N
SMILESOCC1OC(O)C(O)C(O)C1O
さらに表示
Specific optical rotation-49° to -54° (20°C, 589 nm) (c=1, water, NH3)
Appearance (Color)White to almost white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only