Sodium hyaluronate, 95%
Sodium hyaluronate, 95%
Sodium hyaluronate, 95%
Thermo Scientific Chemicals

Sodium hyaluronate, 95%

CAS: 9067-32-7 | (C14H20NO11Na)n | 417.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 251770010
Price (USD)
85.65
Special offer
Online exclusive
Ends: 28-Dec-2026
100.00
Save 14.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
85.65
Special offer
Online exclusive
Ends: 28-Dec-2026
100.00
Save 14.35
Each
Chemical Identifiers
CAS9067-32-7
IUPAC NameSodium hyaluronate
Molecular Formula(C14H20NO11Na)n
SMILESCC(=O)NC1C(OC2C(O)C(O)C(O-*)OC2C(=O)O[Na])OC(CO)C(O)C1O-*
MDL NumberMFCD00875848
SpecificationsSpecification SheetSpecification Sheet
Assay>=94 % (On dry substance)
Protein=<0.2 %
Molecular weight1.5 to 2.2 million Da
Appearance (Form)Powder
Loss on drying=<10 % (500 mg, 105°C, 6 h)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only