Proteinase K Solution (20 mg/mL), RNA grade
Invitrogen™

Proteinase K Solution (20 mg/mL), RNA grade

Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins.Read more
Have Questions?
Catalog NumberQuantity
255300495 mL
Catalog number 25530049
Price (USD)
247.65
Online exclusive
266.00
Save 18.35
Each
Quantity:
5 mL
Price (USD)
247.65
Online exclusive
266.00
Save 18.35
Each
Proteinase K from the fungus Engyodontium album is a nonspecific serine protease that is useful for general digestion of proteins. Proteinase K remains active:

• Over a wide pH range—optimal activity between 6.5 and 9.5
• Under denaturing conditions—e.g., in the presence of SDS or urea
• In the presence of metal chelating agents—e.g., EDTA
• At comparatively high temperatures—optimum digestion temperature is 65°C

Applications
Removal of endogenous nucleases during the preparation of DNA and RNA; preparation of tissue sections for in situ hybridization.

Performance and quality testing
Endodeoxyribonuclease and exodeoxyribonuclease assays; liquid form tested for absence of RNase activity.

Unit definition
One mAnson unit is described as that amount of enzyme that liberates 1 μmole of Folin-positive amino acid within 1 min at 37°C using hemoglobin as a substrate.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Chromatin Biology/RNA Extraction/DNA Extraction
Product TypeProteinase K
PurityDNase-Free, RNase-Free
Quantity5 mL
Shipping ConditionApproved for shipment on Wet or Dry Ice
Concentration20 mg/mL
FormLiquid
Unit SizeEach
Contents & Storage
Store in freezer (-5 to -30°C).

Frequently asked questions (FAQs)

How do you recommend storing proteinase K?

We recommend storing it at 4°C.

How do I isolate DNA from single bacterial colonies before PCR?

Scrape a single colony from the surface of an agar plate and transfer to 12 µL of SCL solution [10 mM Tris-HCl (pH 7.5), 1 mM EDTA, 50 µg/mL proteinase K] and incubate for 15 min at 55 degrees C. Inactivate proteinase K for 15 min at 80 degrees C and add 20 µL of deionized water. Centrifuge at 12,000 x g for 3 min. Transfer the supernatant to a new tube.

What are the recommended conditions for proteinase K treatment when isolating RNA or DNA samples?

Concentration: Generally proteinase K is used in the concentration range of 50 to 500 µg/mL at 65 degrees C in the presence of SDS (0.5-1%).

Temperature optimum: 65 degrees C; 12X more active at 65 degrees C than at 25 degrees C.

pH: Proteinase K is stable over a wide pH range (4.0 to 12.5), with optimal activity at pH 6.5 to 9.5. It is most stable at pH 8.
Inactivation: Heat inactivate Proteinase K at 80 degrees C for 15 min. Phenol extraction is the recommended method to ensure complete inactivation.

Inhibited by: PMSF (0.1 to 1.0 mM of PMSF is usually sufficient for inhibition of Proteinase K)

Not inhibited by: Proteinase K is not inactivated by metal ions, chelating agents (e.g., EDTA), sulfhydryl reagents or by trypsin or chymotrypsin inhibitors. Activity can be stimulated by addition of denaturing agents (SDS and urea).

Citations & References (1)

Citations & References
Abstract
Improved cloning efficiency of polymerase chain reaction (PCR) products after proteinase K digestion.
Authors: Crowe J S; Cooper H J; Smith M A; Sims M J; Parker D; Gewert D;
Journal:Nucleic Acids Res
PubMed ID:2011503
Not available