Search
View additional product information for Pierce™ Crosslink IP Kit - FAQs (26147)
2 product FAQs found
Crosslink IP Kit is covered under our general 1-year warranty and is guaranteed to be fully functional for 12 months from the date of shipment, if stored as recommended. Please see section 8.1 of our Terms & Conditions of Sale (https://www.thermofisher.com/content/dam/LifeTech/Documents/PDFs/Terms-and-Conditions-of-Sale.pdf) for more details.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
No, unfortunately this elution buffer is not sold separately, and we cannot disclose the recipe as it is proprietary.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.