HA Synthetic Peptide - FAQs
HA Synthetic Peptide - FAQs
View additional product information for HA Synthetic Peptide - FAQs (26184)
1 product FAQs found
Once the HA Synthetic Peptide (Cat. No. 26184) is resuspended, at what temperature should I store it?
After resuspending the HA Synthetic Peptide at 1 mg/mL in TBS or water, we recommend aliquoting it and freezing at -20 degrees C. Avoid multiple freeze-thaws.