Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%
Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%
Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%
Thermo Scientific Chemicals

Ethyl cis-2-amino-1-cyclohexanecarboxylate hydrochloride, 95%

CAS: 1127-99-7 | C9H17NO2·HCl | 207.7 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
2.5 g
Packaging:
Glass Bottle
Catalog number 265640025
Price (USD)
343.00
Each
Quantity:
2.5 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
343.00
Each
Chemical Identifiers
CAS1127-99-7
IUPAC Namehydrogen ethyl (1R,2S)-2-aminocyclohexane-1-carboxylate chloride
Molecular FormulaC9H18ClNO2
InChI KeyXMQSOBPCWYVZSW-WLYNEOFISA-N
SMILES[H+].[Cl-].CCOC(=O)[C@@H]1CCCC[C@@H]1N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder and/or chunks
Melting point128°C to 135°C
Titration Argentometric>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only