Unstained mixture of 14 recombinant, highly purified proteins (10kDa to 200kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting.
Have Questions?
Change view
Catalog Number
Quantity
26614
2 x 250 μL
26614X4
8 x 250 μL
2 Options
Catalog number 26614
Price (USD)
113.65
Online exclusive
139.00
Save 25.35
Each
Quantity:
2 x 250 μL
Request bulk or custom format
Price (USD)
113.65
Online exclusive
139.00
Save 25.35
Each
Thermo Scientific PageRuler Unstained Protein Ladder is a mixture of 14 recombinant, highly purified, unstained proteins (10 to 200 kDa) for use as size standards in protein electrophoresis (SDS-PAGE) and western blotting. The protein ladder is supplied in a ready-to-use format for direct loading onto gels; no need to heat, reduce, or add sample buffer prior to use.
Features of the PageRuler Unstained Protein Ladder include:
Reference bands—50 kDa band is stronger than the others for easy orientation
Tagged—the ladder proteins contains an integral Strep-tag™ II sequence and can be detected on western blots using Strep-Tactin™ conjugates or an antibody against Strep-tag™ II Sequence
Applications
Accurate protein sizing on SDS-polyacrylamide gels and western blots
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Gel CompatibilityBolt™ Bis-Tris Plus Gels, Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, NuPAGE™ Tris-Acetate Gels, SDS-PAGE Gels
Shipping ConditionApproved for shipment on Wet or Dry Ice
Number of Markers14
Product LinePageRuler
Product TypeProtein Ladder
Size Range10 to 200 kDa
Stain TypeUnstained
System TypeWestern Blotting, SDS-PAGE
Unit SizeEach
Contents & Storage
Contents: two vials of 250 μL each, proteins at 0.02 to 0.05 mg/mL of each
Storage buffer: 62.5 mM Tris-H3PO4 (pH 7.5 at 25°C), 1 mM EDTA, 2% SDS, 10 mM DTT, 1 mM NaN3, 0.01% bromophenol blue and 33% glycerol
Storage: Upon receipt store at -20°C
Frequently asked questions (FAQs)
The upper bands of the ladder are missing. What could be the reason?
The upper bands of the ladder may be degraded by proteases. Ladder, gel, buffer, pipettes, pipette tips, or equipment can be contaminated by proteases during usage. A general recommendation would be to avoid working with proteases in the same room. We would recommend preparing fresh solutions, cleaning the equipment, and using clean pipettes and tips. If the ladder itself is contaminated, please use a new tube of the ladder.
Some additional bands or smears were observed on the gel when using a PageRuler unstained ladder. What may have caused this?
Additional bands can appear due to dithiothreitol (DTT) oxidation in the storage buffer. Please add newly prepared DTT solution to the final concentration of 100 mM and boil for 5 min at 95 degrees C. This should solve the issue. Addition of DTT is NOT recommended for prestained protein ladders, since too high a concentration of reducing agents can cause protein destaining.
Do the proteins in Thermo Scientific protein ladders have a His-Tag or would otherwise react with an anti-His-Tag antibody?
No, proteins in Thermo Scientific protein ladders are not His tagged. However, non-specific interaction between the ladder proteins and primary or secondary antibodies is possible and some His-Tag detection systems, such as Thermo Scientific 6xHis Protein Tag Stain Reagent Kit, show non-specific interaction. The protein ladder bands are more readily detected when using high antibody concentrations. The non-specific cross-reactivity is difficult to predict, it often has a different pattern dependent on the antibodies used in each individual experiment. The most general way to handle this problem would be to use lower concentrations of antibodies and to use lower amount of protein ladders. It may also be useful to leave one empty well between the ladder and the sample to overcome a possible leakage of the signal to the nearby sample lane.
Can Thermo Scientific protein ladders be detected by Strep-Tactin conjugates?
PageRuler Unstained protein ladders can be detected directly on Western blots by using Strep-Tactin conjugates or an antibody against the Strep-tag II sequence. All PageRuler and Spectra ladder proteins contain an integral Strep-tag II sequence, however the prestained ladders cannot be detected by Strep-Tactin conjugates.
Why is non-specific binding detected after Western blot?
Protein ladder bands can sometimes be detected with chemiluminescent techniques due to non-specific interactions of ladder proteins with either primary or secondary antibodies (or with both). The ladder bands are only rarely detected by chromogenic substrates. The extremely high sensitivity of the chemiluminescent assays is needed to see the bands, so the actual degree of cross-reactivity is low. The non-specific cross-reactivity is difficult to predict, it often has a different pattern depending on the antibodies used. If antibodies recognize a linear epitope, the cross-reactivity may be due to sequence homology. If antibodies react with a denaturation-resistant conformational epitope it could be impossible to identify the exact reason for detected cross-reactivity. The most general way to handle this problem would be to use lower concentrations of antibodies.