Search
View additional product information for Nunc™ 384-Shallow Well Standard Height Polypropylene Sample Processing & Storage Microplates - FAQs (267462, 267461, 267460)
2 product FAQs found
We offer Nunc Microplate Lids: Cat. No. 448143, that fit both the 384-well microplates and the 1,536-well microplates.
We offer Nunc Black or White 384-Well microplates that are intended for use in conjunction with 384-well microplate luminometers and fluorometers that have excitation and detection systems located above the plate. Due to the opaque quality of these microplates, all detection systems must rely on either luminescent or fluorescent reagents, such as viability stains that report cell activity and membrane integrity.