delta-Gluconolactone, 99%
delta-Gluconolactone, 99%
delta-Gluconolactone, 99%
Thermo Scientific Chemicals

delta-Gluconolactone, 99%

CAS: 90-80-2 | C6H10O6 | 178.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number 271050010
Price (USD)
-
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS90-80-2
IUPAC Name3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-one
Molecular FormulaC6H10O6
InChI KeyPHOQVHQSTUBQQK-UHFFFAOYNA-N
SMILESOCC1OC(=O)C(O)C(O)C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Ash=<0.1 %
Appearance (Color)White to off-white
Melting point151°C to 158°C (Dec)
Heavy metals (as Pb)=<20 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only