cis-2-Amino-1-cyclohexanecarboxamide, 98+%, Thermo Scientific Chemicals
cis-2-Amino-1-cyclohexanecarboxamide, 98+%, Thermo Scientific Chemicals
cis-2-Amino-1-cyclohexanecarboxamide, 98+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

cis-2-Amino-1-cyclohexanecarboxamide, 98+%, Thermo Scientific Chemicals

CAS: 115014-77-2 | C7H14N2O | 142.2 g/mol
Have Questions?
Catalog NumberQuantity
2777700101 g
Catalog number 277770010
Price (USD)
-
Quantity:
1 g
Request bulk or custom format
Chemical Identifiers
CAS115014-77-2
IUPAC Name(1S,2R)-2-carbamoylcyclohexan-1-aminium
Molecular FormulaC7H15N2O
InChI KeyDXNKJRKPVQVDJW-RITPCOANSA-O
SMILESNC(=O)[C@@H]1CCCC[C@@H]1[NH3+]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point124°C to 127°C
GC>=98.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only