Search
View additional product information for Nunc™ 96-Well Filter Plates - FAQs (278012, 278011, 278010)
5 product FAQs found
29.936 in. HG equals 1.013 bars
So -5 to -10 in. Hg equals -170 to -340 mbar and -15 to -20 in. Hg equals -500 to -680 mbar
Any vacuum manifold (for 96-well microplates) with an inside depth of 4 cm or more should be fine.
Yes. The polypropylene plate material naturally resists chemicals and organic solvents, as does the polyethylene terephthalate (PET) frit and glass fiber filter. For very strong chemicals, prolonged exposure times, and/or higher temperatures, we recommend that you test the plates with the chemical or solution in question.
The protocol is designed to be flexible, depending on customer needs and instrumentation. For example, certain steps can be performed with vacuum filtration, centrifugation, or positive pressure, with similar results. We do recommend that customers use our formulas for buffers and solutions, since they were optimized to produce superior results.
We recommend using the following Nunc products:
- 96-Well Polypropylene DeepWell Storage Plate, 2 mL (Cat. No. 278743) (sterile growth plate)
- Microplate Lid for 96 microwell plate (Cat. No. 264122) (to cover plate during growth)
- Polyolefin Acrylate Sealing Tape (Cat. No. 232701) (to cover plate during mixing)
- Fritted 96-Well Deep Well Filter Plate (Cat. No. 278011) (for lysate clarification)
- Fritted 96-Well Deep Well Filter Plate with Glass Fiber Filter (Cat. No. 278010) (for DNA binding)
- 96-Well Polypropylene DeepWell Storage Plate, 1.3 mL (Cat. No. 260252) (reception plate)