Brij™-35 Surfact-Amps™ Detergent Solution, 6 x 10 mL - FAQs
Brij™-35 Surfact-Amps™ Detergent Solution, 6 x 10 mL - FAQs
View additional product information for Brij™-35 Surfact-Amps™ Detergent Solution - FAQs (85118, 85117, 28316)
1 product FAQs found
What is the Brij 35 detergent concentration in the 1X Zymogram Developing buffer?
The Brij 35 detergent concentration for Zymogram Developing buffer should be 0.02% (w/v).
Find additional tips, troubleshooting help, and resources within our Protein Electrophoresis and Western Blotting Support Center.