Octylthioglucoside (OTG) Detergent, 5 g - FAQs
Octylthioglucoside (OTG) Detergent, 5 g - FAQs
View additional product information for High-Purity Glucoside Detergents - FAQs (28351, A65515, A65516, 28310)
1 product FAQs found
Are there mass spectrometry reagents for membrane proteins?
CHAPS (Cat. Nos. 28299, 28300), OTG (Cat. No. 28351) and Sodium Deoxycholate (Cat. Nos. 89904, 89905) are detergents often used to solubilize and extract membrane proteins from samples
Find additional tips, troubleshooting help, and resources within our Mass Spectrometry Support Center.