CaptureSelect™ IgG-Fc (Multispecies) Magnetic Agarose Beads
Thermo Scientific™

CaptureSelect™ IgG-Fc (Multispecies) Magnetic Agarose Beads

Thermo Scientific™ CaptureSelect™ IgG-Fc (Multispecies) Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
288285205050 mL
28828520011 mL
28828520055 mL
Catalog number 2882852050
Price (USD)
9,900.00
Each
Quantity:
50 mL
Price (USD)
9,900.00
Each

CaptureSelect IgG-Fc (Multispecies) Magnetic Agarose Beads are high-capacity, high-throughput magnetic affinity particles for antibody purification using manual or robotic magnetic separators. The CaptureSelect IgG-Fc (multispecies) ligand has been specifically designed to purify IgG through high-affinity binding of the IgG Fc region. These magnetic agarose beads can be used for the purification of IgG from a broad range of species. The tested species and their specificity are shown in the figure below.

Features of CaptureSelect IgG-Fc (multispecies) Magnetic Agarose Beads include:

  • One-step IgG purification with high purity, from crude material
  • Compatible with manual and automated workflows (e.g., Thermo Scientific KingFisher instruments)
  • Short operation time
  • No sample dilution required during isolation, repetitive sample loading feasible
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

Main characteristics:

  • Matrix: 6% cross-linked magnetized agarose
  • Average particle size: 25–30 μm
  • Suspension: 25% slurry
  • Ligand: CaptureSelect IgG-Fc (multispecies) affinity ligand
  • Binding capacity: >4 mg IgG/mL slurry
  • Elution conditions: 0.1 M glycine, pH 3.0
Specifications
Binding Capacity>4 mg IgG/mL slurry
Formulation25% (v/v) slurry in Tris buffered saline with 0.05% Tween 20 and 0.05% sodium azide
Ligand TypeCaptureSelect IgG-Fc (multispecies) Affinity Ligand
Matrix6% cross-linked magnetized agarose
Quantity50 mL
Shipping ConditionWet Ice
Sufficient ForSample Preparation
TargetFc-part of IgG of multple species
Capacity (Metric)>4 mg IgG/mL slurry
FormResin Slurry
Particle Size25 to 30 μm
Product LineCaptureSelect
TypeMagnetic Affinity Bead
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C.