CaptureSelect™ CD81 Magnetic Agarose Beads
Thermo Scientific™

CaptureSelect™ CD81 Magnetic Agarose Beads

CaptureSelect CD81 Magnetic Agarose Beads are high-capacity and high-throughput magnetic affinity particles for exosome purification using manual or robotic magnetic separators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
28838420051 x 5 mL
28838420011 x 1 mL
28838420501 x 50 mL
Catalog number 2883842005
Price (USD)
1,724.00
Each
Quantity:
1 x 5 mL
Price (USD)
1,724.00
Each

CaptureSelect CD81 Magnetic Agarose Beads are high-capacity and high-throughput magnetic affinity particles for exosome purification using manual or robotic magnetic separators. The CaptureSelect CD81 affinity ligand recognizes the tetraspanin protein CD81 on the surface of human extracellular vesicles (EVs) like exosomes, enabling purification of CD81-expressing exosomes. CD81-expressing exosomes can be directly isolated from complex source materials in a single step with high purity and yield.

Features of CaptureSelect CD81 Magnetic Agarose Beads include:

  • One-step human CD81-expressing exosome purification with high purity from crude cell clarified supernatant, TFF-concentrated cell lysates, and blood plasma
  • Compatible with manual and automated workflows, e.g., KingFisher instruments
  • Short operation time
  • No sample dilution required during isolation; repetitive sample loading feasible
  • Relatively mild elution at pH 9, resulting in efficient release of exosomes from the magnetic beads.
  • Non-animal-derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12–15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in yeast Saccharomyces cerevisiae by a production process free of any animal components (animal origin-free).

Main characteristics

Matrix: 6% cross-linked magnetized agarose

Average particle size: 25-30 μm

Suspension: 25% slurry

Ligand: CaptureSelect CD81 affinity ligand

Binding capacity: estimated at ≥ 1 x 1010 EVs /mL of magnetic bead slurry

Elution conditions: 50 mM Tris, 1 M arginine, pH 9

Formulation buffer: Tris-buffered saline, 0.05% Tween 20, 0.05 % sodium azide

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Binding Capacity∼1x1010 EVs/mL of Magnetic Beads
Formulation25% (v/v) slurry in Tris buffered saline with 0.05% Tween 20 and 0.05% sodium azide
Ligand TypeCD81 affinity ligand
Matrix6% cross-linked magnetized agarose
Quantity1 x 5 mL
Shipping ConditionWet Ice
Sufficient ForSample Preparation
Capacity (Metric)>1 x 1010 EVs/mL beads
FormResin Slurry
Particle Size25 to 30 μm
Product LineCaptureSelect
TypeMagnetic Agarose Bead
Unit SizeEach
Contents & Storage
Store at 2°C to 8°C