trans-L-3-Hydroxyproline
Thermo Scientific Chemicals

trans-L-3-Hydroxyproline

CAS: 4298-08-2 | C5H9NO3 | 131.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
500 mg
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 297791000
Price (USD)
137.00
Each
Quantity:
100 mg
Request bulk or custom format
Price (USD)
137.00
Each
Chemical Identifiers
CAS4298-08-2
IUPAC Name(2S,3S)-3-hydroxypyrrolidine-2-carboxylic acid
Molecular FormulaC5H9NO3
InChI KeyBJBUEDPLEOHJGE-IMJSIDKUSA-N
SMILESO[C@H]1CCN[C@@H]1C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Total base94.0 to 106.0 %
Appearance (Color)White to yellow to brown
Specific optical rotation-14.5° to -20.5° (20°C, 589 nm) (1% in water)
Water=<2.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only