Search
View additional product information for EZ-Link™ Psoralen-PEG3-Biotin - FAQs (29986)
1 product FAQs found
Yes, the alternative to the Psoralen-Biotin Labeling Kit (Cat. No. AM1480) is the EZ-Link Psoralen-PEG3-Biotin (Cat. No 29986).
Find additional tips, troubleshooting help, and resources within our Labeling Chemistry Support Center.