L-Prolinamide, 98%
L-Prolinamide, 98%
L-Prolinamide, 98%
Thermo Scientific Chemicals

L-Prolinamide, 98%

CAS: 7531-52-4 | C5H11N2O | 115.16 g/mol
Have Questions?
Catalog NumberQuantityPackaging
3029900101 gGlass Bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 302990010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS7531-52-4
IUPAC Name(2S)-2-carbamoylpyrrolidin-1-ium
Molecular FormulaC5H11N2O
InChI KeyVLJNHYLEOZPXFW-BYPYZUCNSA-O
SMILESNC(=O)[C@@H]1CCC[NH2+]1
View more
SpecificationsSpecification SheetSpecification Sheet
HPLC>=97.5 %
Infrared spectrumConforms
Appearance (Form)Powder or crystals
Specific optical rotation-103° to -111° (20°C, 589 nm) (c=1, EtOH)
Appearance (Color)White to light yellow
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only