4-Nitrophenyl-beta-D-glucopyranoside, 99%
4-Nitrophenyl-beta-D-glucopyranoside, 99%
4-Nitrophenyl-beta-D-glucopyranoside, 99%
Thermo Scientific Chemicals

4-Nitrophenyl-beta-D-glucopyranoside, 99%

CAS: 2492-87-7 | C12H15NO8 | 301.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 303110050
Price (USD)
297.65
Special offer
Online exclusive
Ends: 28-Dec-2026
350.00
Save 52.35
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
297.65
Special offer
Online exclusive
Ends: 28-Dec-2026
350.00
Save 52.35
Each
Chemical Identifiers
CAS2492-87-7
IUPAC Name(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(4-nitrophenoxy)oxane-3,4,5-triol
Molecular FormulaC12H15NO8
InChI KeyIFBHRQDFSNCLOZ-RMPHRYRLSA-N
SMILESOC[C@H]1O[C@@H](OC2=CC=C(C=C2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=98.5 %
Specific optical rotation-96° to -117° (20°C, 589 nm) (c=1, water)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only