2-Bromo-4-fluoroaniline, 99%
2-Bromo-4-fluoroaniline, 99%
2-Bromo-4-fluoroaniline, 99%
Thermo Scientific Chemicals

2-Bromo-4-fluoroaniline, 99%

CAS: 1003-98-1 | C6H5BrFN | 190.015 g/mol
Have Questions?
Catalog NumberQuantity
3033100505 g
Catalog number 303310050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1003-98-1
IUPAC Name2-bromo-4-fluoroaniline
Molecular FormulaC6H5BrFN
InChI KeyYLMFXCIATJJKQL-UHFFFAOYSA-N
SMILESNC1=CC=C(F)C=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to light brown
Appearance (Form)Liquid or low melting solid
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only