Nalgene™ Polycarbonate Centrifuge Bottles
Nalgene™ Polycarbonate Centrifuge Bottles
Nalgene™ Polycarbonate Centrifuge Bottles
Nalgene™ Polycarbonate Centrifuge Bottles
Thermo Scientific™

Nalgene™ Polycarbonate Centrifuge Bottles

Process large-volume cell harvesting, pelleting and protein purification with these polycarbonate centrifuge bottles.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNominal CapacityFor Use With (Equipment)Unit Size
3122-0250250 mLFiberlite™ rotorsCase of 36
3122-0250PK
also known as 3122-0250
250 mLFiberlite™ rotorsPack of 4
3122-0500500 mLFiberlite™ rotorsCase of 24
3122-0500PK500 mLFiberlite™ rotorsPack of 4
3122-10001000 mLFiberlite™ rotorsCase of 16
3122-1000PK
also known as 3122-1000
1000 mLFiberlite™ rotorsPack of 4
3122-10101000 mLIEC rotorsCase of 16
3122-1010PK
also known as 3122-1010
1000 mLIEC rotorsPack of 4
Catalog number 3122-0250
Price (USD)
771.15
Online exclusive
1,024.00
Save 252.85
Case of 36
Nominal Capacity:
250 mL
For Use With (Equipment):
Fiberlite™ rotors
Unit Size:
Case of 36
Price (USD)
771.15
Online exclusive
1,024.00
Save 252.85
Case of 36

Process large-volume cell harvesting, pelleting and protein purification with Thermo Scientific™ Nalgene™ Polycarbonate Centrifuge Bottles, the perfect fit with Thermo Scientific™ Fiberlite™ carbon fiber rotors. Molded of transparent polycarbonate, these clear bottles are autoclavable and have excellent mechanical strength properties.

Molded of transparent polycarbonate for easy visualization of samples, these bottles are ideal for low-speed centrifugation of biological and chemical samples.

  • For added leakproof† assurance at maximum speeds or when spinning hazardous materials, use with polypropylene sealing caps assemblies; available for 250mL (DS3131-0038) and 1000mL (DS3132-0063) sizes
  • Cat. No. 3122-1010 designed for use with IEC rotors
  • For proper performance, bottles must be filled to at least 80% of total capacity

Alert:
When spinning a 250mL bottle in a Thermo Scientific Sorvall GSA rotor, the use of the Nalgene centrifuge bottle adaptor (DS3125-0250) is required.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov

† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.

Specifications
AutoclavableAutoclavable
Capacity (English)8.45 oz.
Closure MaterialPolypropylene
Closure TypeScrew Cap
DescriptionPolycarbonate Centrifuge Bottles
Diameter (Metric)61.8 mm
For Use With (Equipment)Fiberlite™ rotors
Height (Metric)125.2 mm
Nominal Capacity250 mL
No. per Case36
No. per Pack4
Product TypeCentrifuge Bottle
ShapeRound
Through the Closure DispensingNo
Capacity (Metric)250 mL
ColorClear
GraduatedNo
MaterialPolycarbonate
Max. Force27,500 x g
Unit SizeCase of 36