EZ-Link™ Plus Activated Peroxidase
EZ-Link™ Plus Activated Peroxidase
EZ-Link™ Plus Activated Peroxidase
Thermo Scientific™

EZ-Link™ Plus Activated Peroxidase

Thermo Scientific Pierce Plus Activated Peroxidase is an amine-reactive form of horseradish peroxidase (HRP) that provides coupling efficiencies of greaterRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
314885 x 1mg
314871 mg
Catalog number 31488
Price (USD)
441.00
Each
Quantity:
5 x 1mg
Request bulk or custom format
Price (USD)
441.00
Each
Thermo Scientific Pierce Plus Activated Peroxidase is an amine-reactive form of horseradish peroxidase (HRP) that provides coupling efficiencies of greater than 95% with antibodies and other proteins. This product consists of 5 mg of lyophilized HRP and is sufficient to modify 5 mg of Immunoglobulin (IgG)

Features of Thermo Scientific Pierce Plus Activated Peroxidase:

• Activated HRP – periodate-treated, aldehyde-activated horseradish peroxidase, ready for conjugation to antibodies and other proteins at sites of primary amines (e.g., lysines)
• Permanent conjugation – reacts efficiently (95%) with primary amines to form covalent amide bonds upon treatment with sodium cyanoborohydride (included in kit)
• High activity HRP – enzyme activity is 120 to 200 units/mg; lyophilized, activated enzyme is stable for at least 12 months at -20°C
• Convenient quantities – each 1 mg-quantity of activated enzyme is sufficient for reaction with 1 mg of IgG to produce about 0.5 mL of conjugate
• Customizable – vary the molar ratios, reaction buffer and pH, and other parameters to acheive conjugates with different levels of HRP incorporation and activity

Plus Activated Peroxidase is horseradish peroxidase (HRP) enzyme, whose native carbohydrates (sugars) have been gently oxidized with periodate to produce amine-reactive aldehyde groups. These carbonyls of Plus Activated Peroxidase spontaneously and efficiently crosslink with primary amines on antibody or other proteins. The method is more effective than other amine-reactive chemistries, such as glutaraldehyde coupling, which often causes polymerization and a greater degree of conjugate inactivation. The pre-activated enzyme eliminates the difficulties inherent in preparing and validating activated peroxidase from scratch. The kit provides the enzyme, accessory reagents and protocol to easily produce high-performance conjugates for Western blotting, ELISA and other detection techniques.

Related Products
EZ-Link™ Plus Activated Peroxidase Kit
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical ReactivityAmine
Label TypeEnzyme Labeled
Label or DyeHRP (Horseradish Peroxidase)
Quantity5 x 1mg
Reactive MoietyAldehyde
SolubilityWater
FormSolid
Product LineEZ-Link
Unit SizeEach
Contents & Storage
Note: EZ-Link Plus Activated Peroxidase contains 1-3 moles of aldehyde per mole of peroxidase.

Storage: Upon receipt store product protected from moisture at -20°C. Product is shipped with an ice pack.

Frequently asked questions (FAQs)

What would be the best way to crosslink an antibody with horseradish peroxidase (‘HRP’ unmodified)?

A commonly used method for conjugating HRP to antibodies is by activation of the HRP using sodium meta-periodate (Cat no. 20504) to generate aldehydes. After the reaction, the HRP must be dialyzed to remove the meta-periodate; then the aldehydes on the HRP will react directly to the amine groups on the antibody. Alternatively, we offer activated HRP (Cat. No. 31488) as an alternative to avoid the periodate oxidation step to modify the HRP. 

Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.

Citations & References (1)

Citations & References
Abstract
A Quantitative Method for Detecting Ara h 2 by Generation and Utilization of Monoclonal Antibodies.
Authors:Chen H, Zou Z, Tao A
Journal:J Immunol Res
PubMed ID:29854839
Peanut (