Methyl-beta-D-galactopyranoside, 98%
Methyl-beta-D-galactopyranoside, 98%
Methyl-beta-D-galactopyranoside, 98%
Thermo Scientific Chemicals

Methyl-beta-D-galactopyranoside, 98%

CAS: 1824-94-8 | C7H14O6 | 194.18 g/mol
数量:
1 g
5 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 314980010
価格(JPY)
-
数量:
1 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1824-94-8
Appearance (Color)White to almost white
HPLC>=97.5 %
Infrared spectrumConforms
Water=<1 % (K.F.)
Appearance (Form)Crystalline powder
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only