Chromosome Medium 1A - FAQs
View additional product information for Chromosome Medium 1A - FAQs (31672017)
1 product FAQs found
Does Chromosome Medium 1A (Cat. No. 31672-017) contain phytohemagglutinin (PHA)?
Yes, Chromosome Medium 1A (Cat. No. 31672-017) contains phytohemagglutinin (PHA) at a concentration of 90 mg/mL. The complete formulation is available here:
https://www.thermofisher.com/us/en/home/technical-resources/media-formulation.342.html