Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • NMDAR1 Antibodies

          Invitrogen

          NMDAR1 Monoclonal Antibody (54.1)

          Advanced Verification
          40 Published Figures
          65 References
          View all (45) NMDAR1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite NMDAR1 Monoclonal Antibody (54.1)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (5)
          • Published Figures (40)
          • Advanced Verification (1)
          NMDAR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          NMDAR1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 46

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          NMDAR1 Antibody (32-0500) in ICC/IF

          Immunofluorescent analysis of MAP2 (green) and NMDAR1 (red) on rat primary Hippocampal neurons (E18) (Product # A15587) cultured for 28 days in the B-27 Plus Neuronal Culture System (Product # A3653401). At day 28 the cells were fixed with 4% paraformaldehyde for 15 min, permeabilized with 0.1% triton x-100 for 30min, and blocked with 1% BSA for 30 min at room temperature. Cells were stained with anti-NMDAR1 antibody (Product # 32-0500) at a dilution of 1:100, and anti-MAP2 (Product # PA5-17646) at a dilution of 1:250, in 1% BSA staining b... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          NMDAR1 Antibody in Immunocytochemistry (ICC/IF)
          NMDAR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Immunoprecipitation (IP)
          NMDAR1 Antibody in Immunocytochemistry (ICC/IF)
          NMDAR1 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          NMDAR1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          NMDAR1 Antibody in Immunohistochemistry (IHC)
          NMDAR1 Antibody in Immunohistochemistry (IHC)
          NMDAR1 Antibody in Immunohistochemistry (IHC)
          NMDAR1 Antibody in Immunohistochemistry (IHC)
          NMDAR1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          NMDAR1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          NMDAR1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody in Western Blot (WB)
          NMDAR1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          32-0500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          View 38 publications 38 publications

          Immunohistochemistry (IHC)

          -
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:50
          View 2 publications 2 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:100
          View 10 publications 10 publications

          ELISA (ELISA)

          0.1-1 µg/mL
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          1-3 µg/mL
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Non-human primate, Rat

          Published species

          Fish, Hamster, Human, Mouse, Rat, Tag

          Host/Isotype

          Mouse / IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          54.1

          Immunogen

          Fusion protein containing sequence from the intracellular loop between transmembrane regions III and IV of NMDAR1.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533060

          Product Specific Information

          This antibody specifically recognizes the ~103 kDa NMDAR1 protein. Cross-reactivity with other NMDA receptor proteins has not been observed.

          Target Information

          NMDAR1 encodes a protein that is a critical subunit of N-methyl-D-aspartate receptors, members of the glutamate receptor channel superfamily which are heteromeric protein complexes with multiple subunits arranged to form a ligand-gated ion channel. These subunits play a key role in the plasticity of synapses, which is believed to underlie memory and learning. Cell-specific factors are thought to control expression of different isoforms, possibly contributing to the functional diversity of the subunits. Alternatively spliced transcript variants have been described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: Glun1r; glutamate [NMDA] receptor subunit zeta-1; glutamate receptor; glutamate receptor, ionotropic, N-methyl D-aspartate 1; N-methyl-D-aspartate glutamate receptor; N-methyl-D-aspartate receptor channel, subunit zeta-1; N-methyl-D-aspartate receptor subunit NR1; neurotransmitter receptor; NMD-R1; NMDA R1 receptor C1 cassette; NMDA receptor 1; NMDZ1; NMZ1; NR1C1; NR1C2; NR1C2'; NR1N1; pGluN1; putative NMDtranscript(altAcc_e2); RP11-350O14.1; unnamed protein product

          View more View less

          Gene Aliases: DEE101; GluN1; GluRdelta1; GluRzeta1; hNR1; M100174; MRD8; NDHMSD; NDHMSR; NMD-R1; NMDA1; Nmdar; NMDAR1; NR1; Rgsc174

          View more View less

          UniProt ID: (Human) Q05586, (Rat) P35439, (Mouse) P35438

          View more View less

          Entrez Gene ID: (Human) 2902, (Rat) 24408, (Mouse) 14810

          View more View less

          Function(s)
          beta-amyloid binding NMDA glutamate receptor activity ion channel activity calcium channel activity calcium ion binding protein binding calmodulin binding ligand-gated ion channel activity ligand-gated sodium channel activity glycine binding glutamate binding transmitter-gated ion channel activity glutamate-gated calcium ion channel activity signaling receptor activity macromolecular complex binding glycine-gated cation channel activity ionotropic glutamate receptor activity receptor binding cation channel activity enzyme binding phosphatase binding voltage-gated cation channel activity glutamate receptor binding ligand-gated monoatomic ion channel activity involved in regulation of presynaptic membrane potential transmitter-gated ion channel activity involved in regulation of postsynaptic membrane potential ionotropic glutamate receptor binding
          Process(es)
          ion transport cation transport chemical synaptic transmission brain development visual learning positive regulation of calcium ion transport into cytosol propylene metabolic process calcium-mediated signaling ionotropic glutamate receptor signaling pathway sodium ion transmembrane transport regulation of membrane potential response to ethanol positive regulation of transcription from RNA polymerase II promoter regulation of synaptic plasticity regulation of neuronal synaptic plasticity protein heterotetramerization positive regulation of synaptic transmission, glutamatergic calcium ion homeostasis excitatory postsynaptic potential calcium ion transmembrane transport calcium ion transmembrane import into cytosol cation transmembrane transport excitatory chemical synaptic transmission positive regulation of reactive oxygen species biosynthetic process regulation of cation transmembrane transport cellular response to beta-amyloid response to glycine positive regulation of excitatory postsynaptic potential conditioned taste aversion response to hypoxia startle response suckling behavior response to amphetamine calcium ion transport cellular calcium ion homeostasis positive regulation of cytosolic calcium ion concentration learning or memory learning memory long-term memory associative learning adult locomotory behavior olfactory learning regulation of cell communication response to amine sensory perception of pain pons maturation hypothalamus development cerebral cortex development social behavior synaptic transmission, glutamatergic positive regulation of apoptotic process response to morphine regulation of neuron apoptotic process negative regulation of neuron apoptotic process regulation of respiratory gaseous exchange regulation of long-term neuronal synaptic plasticity regulation of dendrite morphogenesis regulation of axonogenesis regulation of synapse organization neuromuscular process positive regulation of mitochondrial calcium ion concentration response to calcium ion regulation of synapse assembly regulation of postsynaptic membrane potential prepulse inhibition male mating behavior response to fungicide protein-containing complex assembly cellular response to manganese ion regulation of presynaptic membrane potential chemical synaptic transmission, postsynaptic regulation of plasma membrane bounded cell projection organization positive regulation of Schwann cell migration positive regulation of dendritic spine maintenance response to glycoside protein localization to postsynaptic membrane cellular response to glycine rhythmic process
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-79565875f6-sf8lr:80/100.66.74.151:80.
          git-commit: 280dcbe45276d5d6e613c14d399020b8a49b7116
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.57.0-Offline