Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SRSF1 Antibodies

          Invitrogen

          SRSF1 Monoclonal Antibody (96)

          Advanced Verification
          60 Published Figures
          90 References
          View all (20) SRSF1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SRSF1 Monoclonal Antibody (96)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation'] || 'Additional Information'}}: {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (60)
          • Advanced Verification (2)
          SRSF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SRSF1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 64

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SRSF1 Antibody (32-4500) in WB

          Western blot analysis was performed on whole cell extracts (30 µg lysate) of PC-3 (Lane 1), NTERA-2 (Lane 2), A-431 (Lane 3), MCF7 (Lane 4), Caco-2 (Lane 5), A549 (Lane 6), SH-SY5Y (Lane 7) and HeLa (Lane 8). The blots were probed with Anti-SF2/ASF Mouse Monoclonal Antibody (Product # 32-4500, 2 µg/mL) and detected by chemiluminescence using Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, HRP conjµgate (Product # A28177, 0.4 µg/mL, 1:2500 dilution). A 32 kDa band corresponding to SF2/ASF was observed across the cell lines tested... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Immunocytochemistry, RNA Immunoprecipitation (ICC/IF, RIP)
          SRSF1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          SRSF1 Antibody in Immunocytochemistry (ICC/IF)
          SRSF1 Antibody in Immunocytochemistry (ICC/IF)
          SRSF1 Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          SRSF1 Antibody in Western Blot, Immunohistochemistry (Frozen) (WB, IHC (F))
          SRSF1 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot, ChIP Assay (WB, ChIP)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Western Blot (WB)
          SRSF1 Antibody in Immunoprecipitation (IP)
          SRSF1 Antibody in ChIP Assay (ChIP)
          SRSF1 Antibody in ChIP Assay (ChIP)
          SRSF1 Antibody in ChIP Assay (ChIP)
          SRSF1 Antibody in ChIP Assay (ChIP)
          SRSF1 Antibody in ChIP Assay (ChIP)
          SRSF1 Antibody
          SRSF1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          32-4500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          2 µg/mL
          View 58 publications 58 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          Assay-dependent
          View 4 publications 4 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 10 publications 10 publications

          ChIP assay (ChIP)

          -
          View 7 publications 7 publications

          Dot blot (DB)

          -
          View 1 publication 1 publication

          RNA Immunoprecipitation (RIP)

          -
          View 4 publications 4 publications

          Miscellaneous PubMed (Misc)

          -
          View 4 publications 4 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG2b

          Class

          Monoclonal

          Type

          Antibody

          Clone

          96

          Immunogen

          Recombinant SF2/ASF protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533079

          Target Information

          UBE2E2 catalyzes the covalent attachment of ubiquitin to other proteins.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: alternative; alternative-splicing factor 1; ASF/SF2; OK/SW-cl.3; pre-mRNA-splicing factor SF2, P33 subunit; pre-mRNA-splicing factor SRp30a; splicing factor 2; splicing factor, arginine/serine-rich 1; splicing factor, arginine/serine-rich 1 (ASF/SF2); splicing factor, arginine/serine-rich 1B; splicing factor, arginine/serine-rich, 30-KD, A; SR splicing factor 1; unnamed protein product

          View more View less

          Gene Aliases: 1110054N12Rik; 5730507C05Rik; 6330415C05Rik; ASF; NEDFBA; SF2; SF2p33; SFRS1; SRp30a

          View more View less

          UniProt ID: (Human) Q07955, (Mouse) Q6PDM2

          View more View less

          Entrez Gene ID: (Human) 6426, (Rat) 689890, (Mouse) 110809

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA binding protein binding DNA topoisomerase binding RS domain binding protein kinase B binding
          Process(es)
          alternative mRNA splicing, via spliceosome regulation of alternative mRNA splicing, via spliceosome mRNA 5'-splice site recognition mRNA splicing, via spliceosome mRNA splice site selection mRNA processing positive regulation of RNA splicing protein localization to nucleus regulation of RNA splicing positive regulation of lipid biosynthetic process mRNA stabilization cellular response to interleukin-17 interleukin-17-mediated signaling pathway protein localization to P-body in utero embryonic development RNA splicing oligodendrocyte differentiation cardiac muscle contraction liver regeneration
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-7bf6cff7cf-v8ns5:80/100.66.75.228:80.
          git-commit: cd21911f92a61daaa7859949453b3286702c44a2
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/3.7.0-2026.11.47-1.0