Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • PTBP1 Antibodies

          Invitrogen

          PTBP1 Monoclonal Antibody (1)

          Advanced Verification
          72 Published Figures
          83 References
          View all (26) PTBP1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite PTBP1 Monoclonal Antibody (1)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (6)
          • Published Figures (72)
          • Advanced Verification (1)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 79

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTBP1 Antibody (32-4800) in ICC/IF

          Immunofluorescence analysis of PTB (CLONE 1) was performed using 70% confluent log phase HCT 116 cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 1% BSA for 1 hour at room temperature. The cells were labeled with PTBP1/PTB (1) Mouse Monoclonal Antibody (Product # 32-4800) at 2µg/mL in 0.1% BSA and incubated for 3 hours at room temperature and then labeled with Goat anti-Mouse IgG (H+L) Superclonal™ Secondary Antibody, Alexa Fluor® 488 conjugate (Prod... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Immunoprecipitation (IP)
          PTBP1 Antibody in RNA Immunoprecipitation (RIP)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot, RNA Immunoprecipitation (WB, RIP)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in In Situ Hybridization (ISH)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Immunocytochemistry (ICC/IF)
          PTBP1 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          PTBP1 Antibody in Immunohistochemistry (IHC)
          PTBP1 Antibody in Immunohistochemistry (IHC)
          PTBP1 Antibody in Immunohistochemistry (IHC)
          PTBP1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          PTBP1 Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot, Immunoprecipitation, RNA Immunoprecipitation (WB, IP, RIP)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PTBP1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PTBP1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot, RNA Immunoprecipitation (WB, RIP)
          PTBP1 Antibody in Western Blot, RNA Immunoprecipitation (WB, RIP)
          PTBP1 Antibody in Western Blot, RNA Immunoprecipitation (WB, RIP)
          PTBP1 Antibody in Western Blot, RNA Immunoprecipitation (WB, RIP)
          PTBP1 Antibody in Western Blot, RNA Immunoprecipitation (WB, RIP)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Western Blot (WB)
          PTBP1 Antibody in Immunoprecipitation (IP)
          PTBP1 Antibody in Immunoprecipitation (IP)
          PTBP1 Antibody in Immunoprecipitation (IP)
          PTBP1 Antibody in Immunoprecipitation (IP)
          PTBP1 Antibody in RNA Immunoprecipitation (RIP)
          PTBP1 Antibody in RNA Immunoprecipitation (RIP)
          PTBP1 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          32-4800

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          View 42 publications 42 publications

          Immunohistochemistry (IHC)

          -
          View 3 publications 3 publications

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          2 µg/mL
          View 10 publications 10 publications

          Flow Cytometry (Flow)

          3-5 µg/1x10^6 cells
          View 2 publications 2 publications

          Immunoprecipitation (IP)

          Assay-dependent
          View 12 publications 12 publications

          ChIP assay (ChIP)

          -
          View 2 publications 2 publications

          RNA Immunoprecipitation (RIP)

          Assay-dependent
          View 9 publications 9 publications

          In Situ Hybridization (ISH)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Human, Mouse, Rat, Virus

          Host/Isotype

          Mouse / IgG1

          Class

          Monoclonal

          Type

          Antibody

          Clone

          1

          Immunogen

          Recombinant human PTB protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533082

          Product Specific Information

          This antibody reacts with the C-terminus of the human and rodent PTB protein.

          Target Information

          This gene belongs to the subfamily of ubiquitously expressed heterogeneous nuclear ribonucleoproteins (hnRNPs). The hnRNPs are RNA-binding proteins and they complex with heterogeneous nuclear RNA (hnRNA). These proteins are associated with pre-mRNAs in the nucleus and appear to influence pre-mRNA processing and other aspects of mRNA metabolism and transport. While all of the hnRNPs are present in the nucleus, some seem to shuttle between the nucleus and the cytoplasm. The hnRNP proteins have distinct nucleic acid binding properties. The protein encoded by this gene has four repeats of quasi-RNA recognition motif (RRM) domains that bind RNAs. This protein binds to the intronic polypyrimidine tracts that requires pre-mRNA splicing and acts via the protein degradation ubiquitin-proteasome pathway. It may also promote the binding of U2 snRNP to pre-mRNAs. This protein is localized in the nucleoplasm and it is also detected in the perinucleolar structure. Alternatively spliced transcript variants encoding different isoforms have been described.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: 57 kDa RNA-binding protein PPTB-1; heterogeneous nuclear ribonucleoprotein I; heterogeneous nuclear ribonucleoprotein polypeptide I; hnRNP I; MGC10830; MGC8461; polypyrimidine tract binding protein (heterogeneous nuclear ribonucleoprotein I); pyrimidine binding protein 1; pyrimidine binding protein 2; RNA-binding protein; unnamed protein product

          View more View less

          Gene Aliases: HNRNP-I; HNRNPI; HNRPI; pPTB; PTB; PTB-1; PTB-T; PTB1; PTB2; PTB3; PTB4; Pybp; Pybp1; Pybp2; STADS

          View more View less

          UniProt ID: (Human) P26599, (Rat) Q00438, (Mouse) P17225

          View more View less

          Entrez Gene ID: (Human) 5725, (Rat) 29497, (Mouse) 19205

          View more View less

          Function(s)
          nucleic acid binding RNA binding mRNA binding protein binding poly-pyrimidine tract binding pre-mRNA binding single-stranded DNA endodeoxyribonuclease activity regulatory region RNA binding single-stranded DNA binding sequence-specific DNA binding
          Process(es)
          regulation of alternative mRNA splicing, via spliceosome mRNA processing RNA splicing negative regulation of gene expression negative regulation of RNA splicing regulation of RNA splicing regulation of cell differentiation negative regulation of neuron differentiation negative regulation of mRNA splicing, via spliceosome negative regulation of muscle cell differentiation positive regulation of calcineurin-NFAT signaling cascade IRES-dependent viral translational initiation negative regulation of mRNA metabolic process nuclear-transcribed mRNA catabolic process cardiac ventricle development positive regulation of neuron projection development neurogenesis positive regulation of insulin secretion positive regulation of translation positive regulation of transcription from RNA polymerase II promoter 3'-UTR-mediated mRNA stabilization positive regulation of secretory granule organization
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-frpqk:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch