TMB EASY, Liquid Substrate
TMB EASY, Liquid Substrate
TMB EASY, Liquid Substrate
Thermo Scientific Chemicals

TMB EASY, Liquid Substrate

Have Questions?
Change viewbuttonViewtableView
Substrate Quantity:
30mL
100mL
Catalog number 328031000
Price (USD)
-
Substrate Quantity:
100mL
Request bulk or custom format
Chemical Identifiers
CAS54827-17-7
IUPAC Name3,3',5,5'-tetramethyl-[1,1'-biphenyl]-4,4'-diamine
Molecular FormulaC16H20N2
InChI KeyUAIUNKRWKOVEES-UHFFFAOYSA-N
SMILESCC1=CC(=CC(C)=C1N)C1=CC(C)=C(N)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-green to light blue
Absorbance=<0.05 (450/620 nm, 0.2M H2SO4)
Absorbance(Background level)
Appearance (Form)Solution
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only