n-Dodecyl-beta-D-maltoside, 99%, high purity
n-Dodecyl-beta-D-maltoside, 99%, high purity
n-Dodecyl-beta-D-maltoside, 99%, high purity
Thermo Scientific Chemicals

n-Dodecyl-beta-D-maltoside, 99%, high purity

CAS: 69227-93-6 | C24H46O11 | 510.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Catalog number 329370010
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
181.65
Special offer
Online exclusive
Ends: 28-Dec-2026
214.00
Save 32.35
Each
Chemical Identifiers
CAS69227-93-6
IUPAC Name(2R,3R,4S,5S,6R)-2-{[(2R,3S,4R,5R,6R)-6-(dodecyloxy)-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}-6-(hydroxymethyl)oxane-3,4,5-triol
Molecular FormulaC24H46O11
InChI KeyNLEBIOOXCVAHBD-QKMCSOCLSA-N
SMILESCCCCCCCCCCCCO[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Powder
Specific optical rotation+45° to +49.5° (20°C, 589 nm) (c=1, water)
Infrared spectrumConforms
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only