Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SUMO1 Antibodies

          Invitrogen

          SUMO1 Monoclonal Antibody (21C7)

          68 Published Figures
          159 References
          View all (40) SUMO1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SUMO1 Monoclonal Antibody (21C7)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (68)
          SUMO1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SUMO1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 69

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SUMO1 Antibody (33-2400) in WB

          Western blot analysis of GMP-1 modified RanGAP-1 (90kDa) protein in rat liver nuclei using Ms x GMP-1. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          SUMO1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          SUMO1 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunocytochemistry (ICC/IF)
          SUMO1 Antibody in Immunohistochemistry (IHC)
          SUMO1 Antibody in Immunohistochemistry (IHC)
          SUMO1 Antibody in Western Blot, Immunoprecipitation (WB, IP)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot, Flow Cytometry (WB, Flow)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in Western Blot (WB)
          SUMO1 Antibody in ChIP Assay (ChIP)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          SUMO1 Monoclonal Antibody (21C7)

          Product Details

          33-2400

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-3 µg/mL
          View 80 publications 80 publications

          Immunohistochemistry (IHC)

          Assay-dependent
          View 9 publications 9 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          -
          View 36 publications 36 publications

          Flow Cytometry (Flow)

          -
          View 1 publication 1 publication

          ELISA (ELISA)

          0.1-1.0 µg/mL
          View 1 publication 1 publication

          Immunoprecipitation (IP)

          -
          View 11 publications 11 publications

          ChIP assay (ChIP)

          -
          View 1 publication 1 publication

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication

          Gel Shift (EMSA)

          -
          View 1 publication 1 publication

          Miscellaneous PubMed (Misc)

          -
          View 17 publications 17 publications
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Published species

          Hamster, Human, Mink, Mouse, Non-human primate, Rat, Rhesus monkey, Xenopus

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          21C7

          Immunogen

          Full length recombinant GMP-1
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533109

          Product Specific Information

          This monoclonal antibody can be used to specifically detect the unconjugated (~17 kDa) form of SUMO-1/GMP-1, as well as proteins covalently ligated to GMP-1 (e.g., RanGAP-1). Lysates tested: Rat liver nuclear envelopes, total lysates derived from NIH 3T3 and HeLa cells.

          Target Information

          SUMO1 is an ubiquitin-like protein that can be covalently attached to proteins as a monomer or a lysine-linked polymer. Covalent attachment, via an isopeptide bond, to its substrates requires prior activation by the E1 complex SAE1-SAE2 and linkage to the E2 enzyme UBE2I, and can be promoted by E3 ligases such as PIAS1-4, RANBP2 or CBX4. This post-translational modification on lysine residues of proteins plays a crucial role in a number of cellular processes such as nuclear transport, DNA replication and repair, mitosis and signal transduction. SUMO1 is involved, for instance, in targeting RANGAP1 to the nuclear pore complex protein RANBP2. Polymeric SUMO1 chains are also susceptible to polyubiquitination which functions as a signal for proteasomal degradation of modified proteins. SUMO1 may also regulate a network of genes involved in palate development. Mutations in the gene can result in non-syndromic orofacial cleft 10.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: CG4494-PA; GAP modifying protein 1; LD07775p; Mutations: 288:OK; OTTHUMP00000202403; OTTHUMP00000202404; OTTHUMP00000202405; OTTHUMP00000202406; OTTHUMP00000202409; OTTHUMP00000202410; sentrin; Sentrin-1; similar to ubiquitin and to yeast Smt3p (suppressor of MIF2); Method: conceptual translation supplied by author; SMT3 homolog 3; SMT3 suppressor of mif two 3 homolog 1; SUMO-1; SUMO-1 related peptidase; ubiquitin-homology domain protein PIC1; ubiquitin-like 1; ubiquitin-like 1 (sentrin); ubiquitin-like protein; ubiquitin-like protein SMT3C; ubiquitin-like protein UBL1; UBL1; unnamed protein product

          View more View less

          Gene Aliases: DAP1; GMP1; OFC10; PIC1; SENTRIN; SMT3; SMT3C; SMT3H3; SMTP3; SUMO-1; UBL1

          View more View less

          UniProt ID: (Human) P63165, (Rat) Q5I0H3, (Mouse) P63166

          View more View less

          Entrez Gene ID: (Human) 7341, (Rat) 301442, (Mouse) 22218

          View more View less

          Function(s)
          RNA binding protein binding potassium channel regulator activity enzyme binding protein tag ubiquitin protein ligase binding small protein activating enzyme binding ubiquitin-like protein ligase binding transporter activator activity ubiquitin-specific protease binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter regulation of transcription, DNA-templated protein sumoylation PML body organization positive regulation of protein complex assembly regulation of protein stability positive regulation of proteasomal ubiquitin-dependent protein catabolic process cellular response to heat negative regulation of protein import into nucleus negative regulation of action potential negative regulation of transcription, DNA-templated protein stabilization palate development negative regulation of transcription initiation from RNA polymerase II promoter cellular response to cadmium ion cellular response to hypoxia regulation of cardiac muscle cell contraction protein localization to nuclear pore regulation of calcium ion transmembrane transport DNA repair
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch