cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%
cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%
cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%
Thermo Scientific Chemicals

cis-2-(tert-Butoxycarbonylamino)cyclohexanecarboxylic acid, 98%

CAS: 63216-49-9 | C12H20NO4 | 242.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Catalog number 334270050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS63216-49-9
IUPAC Name(1R,2S)-2-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylate
Molecular FormulaC12H20NO4
InChI KeyQJEQJDJFJWWURK-BDAKNGLRSA-M
SMILESCC(C)(C)OC(=O)N[C@H]1CCCC[C@H]1C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Color)White
Infrared spectrumConforms
Melting point127°C to 131°C
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only