4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%
4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%
4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%
4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%
Thermo Scientific Chemicals

4-Methylumbelliferyl-beta-D-galactopyranoside, 97+%

CAS: 6160-78-7 | C16H18O8 | 338.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 337210050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6160-78-7
IUPAC Name4-methyl-7-{[(2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC16H18O8
InChI KeyYUDPTGPSBJVHCN-DZQJYWQESA-N
SMILESCC1=CC(=O)OC2=CC(O[C@@H]3O[C@H](CO)[C@H](O)[C@H](O)[C@H]3O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to almost white
Specific optical rotation-62° to -66° (20°C, 589 nm) (c=1, pyridine, after 24h)
Infrared spectrumConforms
Water=<1 %
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only