4-Methylumbelliferyl-beta-D-xylopyranoside, 99%
4-Methylumbelliferyl-beta-D-xylopyranoside, 99%
4-Methylumbelliferyl-beta-D-xylopyranoside, 99%
Thermo Scientific Chemicals

4-Methylumbelliferyl-beta-D-xylopyranoside, 99%

CAS: 6734-33-4 | C15H16O7 | 308.29 g/mol
Have Questions?
Catalog NumberQuantity
337272500250 mg
Catalog number 337272500
Price (USD)
393.65
Special offer
Online exclusive
Ends: 28-Dec-2026
463.00
Save 69.35
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
393.65
Special offer
Online exclusive
Ends: 28-Dec-2026
463.00
Save 69.35
Each
Chemical Identifiers
CAS6734-33-4
IUPAC Name4-methyl-7-{[(2S,3R,4S,5R)-3,4,5-trihydroxyoxan-2-yl]oxy}-2H-chromen-2-one
Molecular FormulaC15H16O7
InChI KeyJWIYLOHVJDJZOQ-KAOXEZKKSA-N
SMILESCC1=CC(=O)OC2=CC(O[C@@H]3OC[C@@H](O)[C@H](O)[C@H]3O)=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Infrared spectrumConforms
Appearance of solution(1% in DMF) Almost clear, colorless to
Appearance of solutionlight yellow
HPLC>=98.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only