Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • p38 MAPK beta Antibodies

          Invitrogen

          p38 MAPK beta Monoclonal Antibody (P38-11A5)

          Advanced Verification
          11 Published Figures
          19 References
          View all (20) p38 MAPK beta antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite p38 MAPK beta Monoclonal Antibody (P38-11A5)

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (11)
          • Advanced Verification (2)
          p38 MAPK beta Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          p38 MAPK beta Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 18

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          p38 MAPK beta Antibody (33-8700) in ICC/IF

          Immunofluorescent analysis of p38-beta 2 MAPK Antibody (P38-11A5) was done on 70% confluent log phase HeLa cells. The cells were fixed with 4% paraformaldehyde for 15 minutes, permeabilized with 0.25% Triton™ X-100 for 10 minutes, and blocked with 5% BSA for 1 hour at room temperature. The cells were labeled with p38-beta 2 MAPK Antibody (P38-11A5) (Product # 33-8700) at 1µg/mL in 1% BSA and incubated for 3... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          p38 MAPK beta Antibody in Immunocytochemistry (ICC/IF)
          p38 MAPK beta Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p38 MAPK beta Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p38 MAPK beta Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot, Immunoprecipitation (WB, IP)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody in Western Blot (WB)
          p38 MAPK beta Antibody
          p38 MAPK beta Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          p38 MAPK beta Monoclonal Antibody (P38-11A5)

          Product Details

          33-8700

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 14 publications 14 publications

          Immunohistochemistry (IHC)

          -
          View 1 publication 1 publication

          Immunohistochemistry (Paraffin) (IHC (P))

          1:10-1:100
          -

          Immunocytochemistry (ICC/IF)

          1 µg/mL
          -

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          View 3 publications 3 publications

          Miscellaneous PubMed (Misc)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Human, Mouse, Rabbit

          Host/Isotype

          Mouse / IgG1/IgG2a

          Class

          Monoclonal

          Type

          Antibody

          Clone

          P38-11A5

          Immunogen

          Recombinant, full-length human p38 beta-2; the product of the p38-beta gene
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533144

          Product Specific Information

          33-8700 has been successfully used in immunofluorescence and western blot.

          33-8700 specifically recognizes the beta isoform of p38, with slight reactivity to p38-gamma.

          Target Information

          MAPK11 (p38 beta) is a serine/threonine kinase which is involved in a variety of cellular processes, and is activated by proinflammatory cytokines and environmental stress. Mitogen-activated protein kinase 11. The protein encoded by p38 MAPK beta is a member of the MAP kinase family. MAP kinases act as an integration point for multiple biochemical signals, and are involved in a wide variety of cellular processes such as proliferation, differentiation, transcription regulation, and development. This kinase is most closely related to p38 MAP kinase, both of which can be activated by proinflammatory cytokines and environmental stress. This kinase is activated through its phosphorylation by MAP kinase kinases (MKKs), preferably by MKK6. Transcription factor ATF2/CREB2 has been shown to be a substrate of this kinase.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: MAP kinase 11; MAP kinase p38 beta; MAP kinase p38-2; MAPK 11; MAPK11; mitogen-activated protein kinase p38 beta; mitogen-activated protein kinase p38-2; OTTHUMP00000196655; p38 kinase; protein kinase, mitogen activated kinase, 11, p38beta; splicing isoform of p38Beta MAP Kinase; stress-activated protein kinase-2; stress-activated protein kinase-2b; unnamed protein product

          View more View less

          Gene Aliases: p38-2; P38B; p38Beta; P38BETA2; PRKM11; SAPK2; SAPK2B

          View more View less

          UniProt ID: (Human) Q15759, (Mouse) Q9WUI1

          View more View less

          Entrez Gene ID: (Human) 5600, (Mouse) 19094

          View more View less

          Function(s)
          protein kinase activity protein serine/threonine kinase activity MAP kinase activity protein binding ATP binding protein serine kinase activity
          Process(es)
          MAPK cascade osteoblast differentiation positive regulation of gene expression stress-activated protein kinase signaling cascade positive regulation of interleukin-12 production intracellular signal transduction p38MAPK cascade positive regulation of erythrocyte differentiation positive regulation of muscle cell differentiation stress-activated MAPK cascade bone development cellular response to interleukin-1 cellular response to UV-B cellular senescence cellular response to virus protein phosphorylation protein maturation regulation of cardiac muscle cell proliferation negative regulation of cardiac muscle cell proliferation pyroptosis NLRP1 inflammasome complex assembly
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-q86qf:80/100.66.76.64:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch