Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          • Primary Antibodies ›
          • Ankyrin G Antibodies

          Invitrogen

          Ankyrin G Monoclonal Antibody (4G3F8)

          Advanced Verification
          27 Published Figures
          46 References
          View all (26) Ankyrin G antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Tech Support

          Cite Ankyrin G Monoclonal Antibody (4G3F8)

          Additional Information:

          {{$ctrl.isSkuNotesCollapsed ? 'View more' : 'View less'}}
          • Antibody Testing Data (3)
          • Published Figures (27)
          • Advanced Verification (1)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 31

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Ankyrin G Antibody (33-8800) in ICC/IF

          Immunofluorescence analysis of Ankyrin G was performed using 70% confluent log phase SH-SY5Y cells. The cells were fixed with 4% paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 15 minutes, and blocked with 2% BSA for 45 minutes at room temperature. The cells were labeled with Ankyrin G Monoclonal Antibody (4G3F8) (Product # 33-8800) at 5 µg/mL in 0.1% BSA, incubated at 4 degree ce... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Western Blot (WB)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunocytochemistry (ICC/IF)
          Ankyrin G Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Ankyrin G Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Ankyrin G Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Ankyrin G Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Ankyrin G Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Ankyrin G Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Ankyrin G Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Ankyrin G Antibody in Immunohistochemistry (IHC)
          Ankyrin G Antibody in Immunohistochemistry (IHC)
          Ankyrin G Antibody in Immunohistochemistry (IHC)
          Ankyrin G Antibody in Immunohistochemistry (PFA fixed) (IHC (PFA))
          Ankyrin G Antibody in Western Blot (WB)
          Ankyrin G Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          33-8800

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 2 publications 2 publications

          Immunohistochemistry (IHC)

          -
          View 11 publications 11 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          -
          View 1 publication 1 publication

          Immunohistochemistry (PFA fixed) (IHC (PFA))

          -
          View 2 publications 2 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 4 publications 4 publications

          Immunocytochemistry (ICC/IF)

          2-5 µg/mL
          View 25 publications 25 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Proximity Ligation Assay (PLA) (PLA)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Dog, Human, Mouse, Rabbit, Rat

          Published species

          Bovine, Dog, Fruit fly, Hamster, Human, Mouse, Pig, Rat, Sheep, Xenopus

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          4G3F8

          Immunogen

          A synthetic peptide derived from the spectrin-binding domain of the human ankyrin G protein.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533145

          Target Information

          Ankyrin-G is found in several tissues including brain where, due to tissue-specific RNA processing, it is expressed as two different isoforms, localized to axon initial segments and nodes of Ranvier. Ankyrin-G is also found at sites of epithelial cell-cell contact and in the intracellular membranes of some cells. Ankyrin-G isoforms contain a unique stretch of sequence highly enriched in serine and threonine residues immediately following a globular head domain. Ankyrin-G is thought to participate in the maintenance and targeting of ion channels and cell adhesion molecules to the nodes of Ranvier and to axon initial segments.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: ANK-3; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin 3, node of Ranvier (ankyrin G); ankyrin G; ankyrin-G; brain-specific ankyrin-G; cytoskeletal protein; unnamed protein product

          View more View less

          Gene Aliases: 2900054D09Rik; ANK-3; AnkG; Ankyrin-3; ANKYRIN-G; MRT37

          View more View less

          UniProt ID: (Human) Q12955, (Mouse) G5E8K5, (Rat) O70511

          View more View less

          Entrez Gene ID: (Human) 288, (Rabbit) 100353296, (Dog) 479215, (Mouse) 11735, (Rat) 361833

          View more View less

          Function(s)
          structural constituent of cytoskeleton protein binding cytoskeletal protein binding cytoskeletal adaptor activity sodium channel regulator activity spectrin binding protein binding, bridging ion channel binding cadherin binding channel activator activity phosphorylation-dependent protein binding RNA polymerase II sequence-specific DNA binding transcription factor binding scaffold/adaptor protein
          Process(es)
          mitotic cytokinesis plasma membrane organization cytoskeleton organization signal transduction axonogenesis neuromuscular junction development positive regulation of gene expression positive regulation of cell communication by electrical coupling positive regulation of sodium ion transport magnesium ion homeostasis neuronal action potential positive regulation of homotypic cell-cell adhesion Golgi to plasma membrane protein transport regulation of potassium ion transport establishment of protein localization positive regulation of membrane potential Golgi vesicle transport cellular response to magnesium ion membrane assembly protein localization to plasma membrane maintenance of protein location in plasma membrane positive regulation of protein targeting to membrane protein localization to axon positive regulation of membrane depolarization during cardiac muscle cell action potential negative regulation of delayed rectifier potassium channel activity axon guidance anterograde axonal transport establishment or maintenance of microtubule cytoskeleton polarity maintenance of protein location in cell clustering of voltage-gated sodium channels positive regulation of action potential negative regulation of endocytosis synapse organization axon development inorganic ion homeostasis regulation of action potential regulation of modification of postsynaptic structure positive regulation of NIK/NF-kappaB signaling regulation of protein targeting positive regulation of sodium ion import across plasma membrane regulation of cation transmembrane transport regulation of transcription from RNA polymerase II promoter response to immobilization stress
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-5bc99cc877-lxljx:80/100.66.74.151:80.
          git-commit: a70b05477dc28cec55d7eafa2ce360191b99e591
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.58.0-2026.09.99-1.0