Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • SR Antibodies

          Invitrogen

          SR Monoclonal Antibody (16H3 (16H3E8))

          3 Published Figures
          19 References
          View all (3) SR antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite SR Monoclonal Antibody (16H3 (16H3E8))

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (1)
          • Published Figures (3)
          SR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          SR Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          SR Antibody (33-9300) in WB

          Western blot analysis of SR proteins expression in HeLa cell lysate using SR Protein Monoclonal Antibody, Mouse {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          SR Antibody in Western Blot (WB)
          SR Antibody in Western Blot (WB)
          SR Antibody in Western Blot (WB)
          SR Antibody in Western Blot (WB)

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Product Details

          33-9300

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          3-5 µg/mL
          View 12 publications 12 publications

          Immunocytochemistry (ICC/IF)

          0.5-2 µg/mL
          View 2 publications 2 publications

          ELISA (ELISA)

          0.1-1 µg/mL
          -

          Immunoprecipitation (IP)

          -
          View 2 publications 2 publications

          Miscellaneous PubMed (Misc)

          -
          View 3 publications 3 publications
          Product Specifications

          Species Reactivity

          Bovine, Dog, Chicken, Fruit fly, Human, Mouse, Rabbit, Rat, Xenopus

          Published species

          Human, Mouse, Rat

          Host/Isotype

          Mouse / IgG1, kappa

          Class

          Monoclonal

          Type

          Antibody

          Clone

          16H3 (16H3E8)

          Immunogen

          Purified, de-phosphorylated bovine Srp55 and Drosophila p55-GST. The 16H3 epitope has been mapped to a 40-amino acid polypeptide composed almost exclusively of arginine alternating with glutamate and aspartate

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Amount

          100 µg

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533149

          Product Specific Information

          This monoclonal antibody can be used to detect a sub-set of the non-snRNP splicing factors termed SR proteins. The antibody detects SRp75, SRp55, SRp40, and SRp20 but not SRp30a or b (ASF/SF2, SC-35) proteins. 16H3 also recognizes approximately 20 distinct nuclear proteins including the U1 70K component of the U1 snRNP and both subunits of U2AF. The epitope detected by the 16H3 monoclonal antibody has been termed the 'alternating arginine domain' and is composed almost exclusively of argnine alternating with glutamate and aspartate. 16H3 targets are predominantly nuclear, nonnucleolar, and are localized to active sites of polymerase II transcription.

          Target Information

          SR proteins are a highly conserved family of arginine/serine-rich, spliceosome-associated phosphoproteins essential for metazoan pre-mRNA splicing. SR proteins act early in splicing by promoting splice site recognition and spliceosome assembly. SR proteins also play a regulatory role because they can determine alternative splice site usage in vivo and in vitro. SR proteins appear to be recruited from nuclear 'speckles', in which they are concentrated to sites of transcription in order to spatially coordinate transcription and pre-mRNA splicing within the cell nucleus.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: delayed-early protein HRS; epididymis secretory protein Li 91; epididymis secretory sperm binding protein; insulin-induced growth response protein CL-4; pre-mRNA splicing factor SRp20; pre-mRNA-splicing factor SRP20; pre-mRNA-splicing factor SRP40; pre-mRNA-splicing factor SRP55; pre-mRNA-splicing factor SRP75; putative; RNA binding protein; serine/arginine-rich splicing factor 4; SFRS5; splicing factor SRp55; splicing factor SRp55-1; splicing factor, arginine/serine-rich 3; splicing factor, arginine/serine-rich 3 (SRp20); splicing factor, arginine/serine-rich 4; splicing factor, arginine/serine-rich 4 (SRp75); splicing factor, arginine/serine-rich 5; splicing factor, arginine/serine-rich 5 (SRp40, HRS); splicing factor, arginine/serine-rich 6; splicing factor, arginine/serine-rich, 20-kD; splicing factor, arginine/serine-rich, 55 kDa; SR splicing factor 4; SR splicing factor 5; SR splicing factor 6; SRP001LB; unnamed protein product; X16

          View more View less

          Gene Aliases: 1210001E11Rik; 5730499P16Rik; B52; HEL-S-91; HRS; MNCb-2616; RGD1561347; SFRS3; SFRS4; SFRS5; SFRS6; SRP20; SRP40; SRP55; SRP55-1; SRP75; X16

          View more View less

          UniProt ID: (Bovine) Q3SZR8, (Human) P84103, (Human) Q08170, (Human) Q13243, (Human) Q13247, (Bovine) Q3B7L6, (Mouse) P84104, (Rat) Q09167, (Mouse) O35326, (Mouse) Q3TWW8, (Rat) G3V6S8

          View more View less

          Entrez Gene ID: (Dog) 403687, (Chicken) 419815, (Bovine) 540276, (Human) 6428, (Rabbit) 100347748, (Rabbit) 100350179, (Bovine) 614310, (Chicken) 419600, (Dog) 478160, (Human) 6429, (Rabbit) 100340192, (Dog) 490754, (Chicken) 423258, (Human) 6430, (Bovine) 510474, (Human) 6431, (Chicken) 419116, (Bovine) 507828, (Dog) 403667, (Rabbit) 100008654, (Mouse) 20383, (Rat) 361814, (Rat) 362612, (Mouse) 57317, (Rat) 29667, (Mouse) 20384, (Mouse) 67996, (Rat) 362264

          View more View less

          Function(s)
          RNA binding phospholipase binding nucleic acid binding mRNA binding protein binding pre-mRNA binding primary miRNA binding protein-RNA sequence-specific adaptor activity sequence-specific mRNA binding protein kinase B binding RS domain binding RNA splicing factor
          Process(es)
          mRNA export from nucleus regulation of mRNA splicing, via spliceosome cellular response to leukemia inhibitory factor regulation of alternative mRNA splicing, via spliceosome RNA processing primary miRNA processing RNA splicing, via transesterification reactions mRNA processing RNA splicing response to insulin negative regulation of mRNA splicing, via spliceosome liver development mRNA splice site selection cellular response to insulin stimulus positive regulation of RNA splicing liver regeneration alternative mRNA splicing, via spliceosome negative regulation of gene expression regulation of keratinocyte proliferation negative regulation of keratinocyte differentiation positive regulation of epithelial cell proliferation involved in lung morphogenesis regulation of wound healing negative regulation of type B pancreatic cell apoptotic process insulin receptor signaling pathway biological_process hematopoietic progenitor cell differentiation response to wounding
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-7kv6q:80/100.66.72.150:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch