1-Step™ TMB ELISA Substrate Solutions
Thermo Scientific™

1-Step™ TMB ELISA Substrate Solutions

Thermo Scientific™ Pierce TMB ELISA Substrates detect horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantitySubstrate
34021400 mL KitTMB Substrate
34024250 mLSlow TMB-ELISA Substrate
34022250 mLTurbo TMB-ELISA Substrate
34028250 mLUltra TMB-ELISA Substrate
340291 LUltra TMB-ELISA Substrate
Catalog number 34021
Price (USD)
205.65
Online exclusive
231.00
Save 25.35
Each
Quantity:
400 mL Kit
Substrate:
TMB Substrate
Request bulk or custom format
Price (USD)
205.65
Online exclusive
231.00
Save 25.35
Each

TMB (3,3',5,5' tetramethylbenzidine) is a chromagen that yields a blue color when oxidized with hydrogen peroxide (catalyzed by HRP) with major absorbances at 370 nm and 652 nm. The color then changes to yellow (measured at 450 nm) upon addition of sulfuric or phosphoric acid to stop the reaction. TMB is very sensitive and more quickly oxidized than other HRP substrates, resulting in faster color development. Unlike other commercially available substrates, these products contain no DMF or DMSO.

There are three formulations that differ primarily in their sensitivities. Slow TMB is intermediate in sensitivity, ideal for kinetic readings. The sensitivity of the Turbo TMB compares to that of OPD used at approximately 1 mg/mL. Ultra TMB-ELISA produces the highest signal-to-noise ratio and sensitivity in the picogram range.

Thermo Scientific Pierce 1-Step TMB ELISA Substrates detects horseradish peroxidase (HRP) activity, yielding a blue color (Amax = 370 nm and 652 nm) that changes to yellow (Amax = 450 nm) upon addition of a sulfuric or phosphoric acid stop solution.

Features of 1-Step TMB ELISA Substrates:
• Available in traditional or ready-to-use formulations
• No additional reagents or filtering required
• Non carcinogenic
• Various levels of sensitivity and development speed to suit any assay
• No DMF or DMSO present in the reagents

TMB is the most popular chromogenic substrate for HRP detection in ELISA and is available in several formats. The Pierce 1-Step TMB Substrates are one-component substrates that require no preparation before use. Unlike other commercially available substrates, these products contain no DMF or DMSO.

Thermo Scientific Pierce 1-Step Ultra TMB yields the greatest sensitivity among the TMB substrates, followed by 1-Step Turbo TMB and 1-Step Slow TMB. The sensitivity of the 1-Step Turbo TMB is similar to OPD-based substrates formulated at approximately 1 mg/mL. 1-Step Slow TMB has intermediate sensitivity—more sensitive than ABTS, but less sensitive than o-phenylenediamine dihydrochloride (OPD) or 1-Step Turbo TMB. The 1-Step Slow TMB is an ideal substrate for kinetic studies.

TMB (3,3',5,5'-tetramethylbenzidine) is a chromogen that yields a blue color when oxidized, typically as a result of oxygen radicals produced by the hydrolysis of hydrogen peroxide by HRP. For kinetic or non-stopped ELISA assays, the TMB chromogen has maximal absorbances at 370 nm and 652 nm. The color then changes to yellow with the addition of sulfuric or phosphoric acid with maximum absorbance at 450 nm. A green reaction product may result from partial conversion to the yellow product from the blue intermediate. TMB is very sensitive ELISA substrate and is more quickly oxidized than other HRP substrates, resulting in faster color development.

For Research Use Only. Not for use in diagnostic procedures.

1-Step Substrates are available in three formulations that vary in sensitivity:

  • 1-Step Slow TMB has intermediate sensitivity – more sensitive than ABTS, but less sensitive than OPD or 1-Step Turbo TMB
  • 1-Step Turbo TMB has sensitivity comparable to OPD used at approximately 1mg/mL
  • 1-Step Ultra TMB-ELISA has the highest signal-to-noise ratio and sensitivity in the picogram range

General References:

  1. Josephy, P.D., et al. (1982). J. Biol. Chem. 257(7), 3669-3675.
  2. Josephy, P.D., et al. (1983). J. Biol. Chem. 258(9), 5561-5569.
  3. Liem, H.H., et al. (1979). Anal. Biochem. 98, 388-393.
Specifications
Detection MethodColorimetric
Product LinePierce
SubstrateTMB Substrate
Substrate PropertiesChemical Substrate
Substrate TypeHRP (Horseradish Peroxidase)
Product TypeELISA Substrate
Quantity400 mL Kit
Unit SizeEach
Contents & Storage
Sufficient for: 400 mL of typical 1X TMB buffer (4000 wells at 100 μL per well)
• TMB Solution, 200 mL
• Peroxide Solution, 200 mL

Store at 4°C.

Frequently asked questions (FAQs)

What substrate solution do you recommend using with IL-8 Human Matched Antibody Pair (Cat. No. MS204-3MST)?

We recommend the following formulation when making your own: 1:2 mixture of H2O2 and tetramethylbenzidine.
However we also offer a few alternatives for purchase:
- Pierce TMB Substrate Kit (Cat. No. 34021)
- Support Pack Standard (Cat. No. BMS412) and Support Pack Plus (Cat. No. BMS414). Both of these ELISA Support Packs include stabilized TMB substrate (does not require mixing), assay buffer, wash buffer, stop solution and PBS. Support Pack Plus includes plates.

Find additional tips, troubleshooting help, and resources within our Antibodies and Immunoassays Support Center.