Nalgene™ Natural PPCO Micro Packaging Vials: Sterile, Bulk Pack, 1.5mL - FAQs
View additional product information for Nalgene™ Natural PPCO Micro Packaging Vials: Sterile, Bulk Pack - FAQs (342800-0015, 342800-0005, 342800-0045, 342800-0020)
1 product FAQs found
Are the Nalgene PPCO media bottles/tubes able to be irradiated?
PPCO bottles/tubes are not recommended for sterilization by radiation. Polycarbonate bottles/tubes would be a suitable alternative if radiation sterilization is desired.