D-Threonine, 98%
D-Threonine, 98%
D-Threonine, 98%
Thermo Scientific Chemicals

D-Threonine, 98%

CAS: 632-20-2 | C4H9NO3 | 119.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
Catalog number 346970050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS632-20-2
IUPAC Name(2R,3S)-2-amino-3-hydroxybutanoic acid
Molecular FormulaC4H9NO3
InChI KeyAYFVYJQAPQTCCC-STHAYSLISA-N
SMILESC[C@H](O)[C@@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals
Chloride (Cl)=<200 ppm
Sulfate (SO4)=<300 ppm
Infrared spectrumConforms
Loss on drying=<0.3 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only