(1S,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee
(1S,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee
(1S,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee
Thermo Scientific Chemicals

(1S,3R)-N-BOC-1-Aminocyclopentane-3-carboxylic acid, 95%, 98% ee

CAS: 161660-94-2 | C11H18NO4 | 228.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Packaging:
Glass Bottle
Catalog number 361842500
Price (USD)
150.00
Each
Quantity:
250 mg
Packaging:
Glass Bottle
Request bulk or custom format
Price (USD)
150.00
Each
Chemical Identifiers
CAS161660-94-2
IUPAC Name(1R,3S)-3-{[(tert-butoxy)carbonyl]amino}cyclopentane-1-carboxylate
Molecular FormulaC11H18NO4
InChI KeyRNJQBGXOSAQQDG-SFYZADRCSA-M
SMILESCC(C)(C)OC(=O)N[C@H]1CC[C@H](C1)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Enantiomeric excess>=97.5 %
Appearance (Form)Powder
HPLC>=94 %
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only