Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Claudin 11 Antibodies

          Invitrogen

          Claudin 11 Polyclonal Antibody

          Advanced Verification
          40 Published Figures
          100 References
          View all (9) Claudin 11 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Claudin 11 Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (2)
          • Published Figures (40)
          • Advanced Verification (2)
          Claudin 11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Claudin 11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 44

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Claudin 11 Antibody (36-4500) in IHC (P)

          Immunohistochemistry analysis of Claudin-11 showing staining in the membrane of paraffin-embedded mouse brain tissue (right) compared to a negative control without primary antibody (left). To expose target proteins, antigen retrieval was performed using 10mM sodium citrate (pH 6.0), microwaved for 8-15 min. Following antigen retrieval, tissues were blocked in 3% H2O2-methanol for 15 min at room temperature,... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Claudin 11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Western Blot, Immunohistochemistry (WB, IHC)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Claudin 11 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 11 Antibody in Immunohistochemistry, Immunocytochemistry (IHC, ICC/IF)
          Claudin 11 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 11 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 11 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 11 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 11 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 11 Antibody in Western Blot, Immunocytochemistry (WB, ICC/IF)
          Claudin 11 Antibody in Immunocytochemistry (ICC/IF)
          Claudin 11 Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Claudin 11 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 11 Antibody in Immunohistochemistry (Frozen) (IHC (F))
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Immunohistochemistry (IHC)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody in Western Blot (WB)
          Claudin 11 Antibody
          Claudin 11 Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Claudin 11 Polyclonal Antibody

          Product Details

          36-4500

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          View 32 publications 32 publications

          Immunohistochemistry (IHC)

          -
          View 33 publications 33 publications

          Immunohistochemistry (Paraffin) (IHC (P))

          1:20-1:200
          View 5 publications 5 publications

          Immunohistochemistry (Frozen) (IHC (F))

          -
          View 7 publications 7 publications

          Immunocytochemistry (ICC/IF)

          -
          View 21 publications 21 publications

          ELISA (ELISA)

          Assay-dependent
          -

          Immunoprecipitation (IP)

          Assay-dependent
          -

          Miscellaneous PubMed (Misc)

          -
          View 2 publications 2 publications
          Product Specifications

          Species Reactivity

          Human, Mouse

          Published species

          Cynomolgus monkey, Goat, Hamster, Human, Mouse, Pig, Plant, Rabbit, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic peptide derived from the C-terminal region of human OSP/Claudin-11, which differs by only one amino acid from mouse and rat sequences.
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Amount

          100 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          0.1% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2533259

          Target Information

          The claudin superfamily consists of many structurally related proteins in humans. These proteins, which include claudin 1 through 18, are important structural and functional components of tight junctions in paracellular transport. Claudins are located in both epithelial and endothelial cells in all tight junction-bearing tissues. Claudins, which consist of four trans-membrane domains and two extracellular loops, make up tight junction strands. Claudin-11 is an oligodendrocyte-specific protein that is expressed in the tight junctions of Sertoli cells and myelin sheaths in mice.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: oligodendrocyte transmembrane protein; oligodendrocyte-specific protein; unnamed protein product

          View more View less

          Gene Aliases: Claudin-11; Claudin11; HLD22; OSP; OTM

          View more View less

          UniProt ID: (Human) O75508, (Mouse) Q60771

          View more View less

          Entrez Gene ID: (Human) 5010, (Mouse) 18417

          View more View less

          Function(s)
          structural molecule activity protein binding identical protein binding
          Process(es)
          cell adhesion calcium-independent cell-cell adhesion via plasma membrane cell-adhesion molecules tight junction assembly spermatogenesis axon ensheathment
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch