Search
View additional product information for Clear Round-Bottom Immuno Nonsterile 96-Well Plates - FAQs (449824, 475434, 3555TS, 3655TS)
1 product FAQs found
For energy treated plates, the entire plate surface receives the energy treatment. The MaxiSorp, PolySorp, MediSorp and MultiSorp plates are energy treated plates. For traditionally treated plates, the well surface is treated up to the listed working volume.