Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • New Products
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          • Primary Antibodies ›
          • Tyrosine Hydroxylase Antibodies

          Invitrogen

          Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal Antibody

          Advanced Verification
          4 Published Figures
          11 References
          View all (61) Tyrosine Hydroxylase antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers
          Datasheet
          Certificates
          Protocols
          Questions & Answers

          Cite Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal Antibody

          {{$ctrl.translations['antibody.pdp.skuNotes.additionalInformation']}}:

          {{$ctrl.translations[$ctrl.isSkuNotesCollapsed ? 'antibody.pdp.skuNotes.viewMore' : 'antibody.pdp.skuNotes.viewLess']}}
          • Antibody Testing Data (5)
          • Published Figures (4)
          • Advanced Verification (3)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 12

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Tyrosine Hydroxylase (Ser40) Antibody (36-8600) in ICC/IF

          Immunofluorescence analysis of Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal Antibody was performed using 70% confluent log phase PC-12 cells. The cells were fixed with 4% Paraformaldehyde for 10 minutes, permeabilized with 0.1% Triton™ X-100 for 10 minutes, and blocked with 2% BSA for 10 minutes at room temperature. The cells were labeled with Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal Antibody (Pro... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Immunohistochemistry (IHC)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Western Blot (WB)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Western Blot (WB)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Immunohistochemistry (IHC)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Western Blot (WB)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Western Blot (WB)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody in Western Blot (WB)
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody
          Phospho-Tyrosine Hydroxylase (Ser40) Antibody

          Please note: We are reviewing Western blot images included in the antibody testing data in our catalog, including those provided by third parties. Unless expressly labeled or annotated as “raw-unedited”, Western blot images included in the antibody testing data in our catalog may have been edited, optimized or otherwise adjusted for presentation.

          Phospho-Tyrosine Hydroxylase (Ser40) Polyclonal Antibody

          Product Details

          36-8600

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          View 7 publications 7 publications

          Immunohistochemistry (IHC)

          1:1,000
          View 3 publications 3 publications

          Immunohistochemistry - Free Floating (IHC (Free))

          -
          View 1 publication 1 publication

          Immunocytochemistry (ICC/IF)

          1:1,000
          -

          ELISA (ELISA)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Rat

          Published species

          Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Synthetic phospho-peptide derived from the rat tyrosine hydroxylase (TH) protein (aa 40).
          3D Epitope / Immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.3 mg/mL

          Amount

          30 µg

          Purification

          Antigen affinity chromatography

          Storage buffer

          0.01M HEPES, pH 7.5, with 50% glycerol, 100µg/mL BSA, 0.15M NaCl

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_138590

          Target Information

          Tyrosine hydroxylase (TH) is an enzyme involved in the synthesis of catecholamine neurotransmitters dopamine, epinephrine, and norepinephrine. In all species, catecholamine synthesis is regulated by the interaction of TH with a cofactor, tetrahydrobiopterin (BH4). BH4 binds to the TH catalytic domain, resulting in enzymatic activity. Unlike TH in non-primate species, four human TH mRNA splice variants (hTH1-hTH4) have been isolated. These variants are identical in their catalytic domain, but differ in their N-terminal, regulatory domains. TH is also responsible for the conversion of L-tyrosine to L-dopa. TH plays a key role in the physiology of adrenergic neurons. The role of TH in the synthesis of catecholamine neurotransmitters suggests a correlation between the enzyme and a number of neuropathogenic diseases including: Parkinson's disease, schizophrenia, Segawa syndrome, and dystonia, as well as a variety of cardiovascular diseases.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: EC 1.14.16.2; HGNC:11782; TH isoform 3; TH isoform a; TH-4; TY3H; TYH antibody; tyrosine 3-hydroxylase; tyrosine hydroxylase (EC 1.14.16.2)

          View more View less

          Gene Aliases: The

          View more View less

          UniProt ID: (Rat) P04177

          View more View less

          Entrez Gene ID: (Rat) 25085

          View more View less

          Function(s)
          monooxygenase activity tyrosine 3-monooxygenase activity iron ion binding protein binding ferrous iron binding ferric iron binding amino acid binding oxidoreductase activity, acting on paired donors, with incorporation or reduction of molecular oxygen, reduced pteridine as one donor, and incorporation of one atom of oxygen oxygen binding enzyme binding protein domain specific binding tetrahydrobiopterin binding dopamine binding small molecule binding identical protein binding
          Process(es)
          response to hypoxia synaptic transmission, dopaminergic response to amphetamine dopamine biosynthetic process from tyrosine fatty acid metabolic process sphingolipid metabolic process heart development visual perception learning memory mating behavior locomotory behavior regulation of heart contraction aromatic amino acid family metabolic process response to xenobiotic stimulus response to light stimulus response to herbicide response to salt stress organ morphogenesis response to metal ion response to zinc ion response to activity aminergic neurotransmitter loading into synaptic vesicle response to insecticide phthalate metabolic process cerebral cortex development response to nutrient levels response to estradiol response to lipopolysaccharide response to nicotine social behavior response to isolation stress response to immobilization stress dopamine biosynthetic process epinephrine biosynthetic process norepinephrine biosynthetic process catecholamine biosynthetic process eye photoreceptor cell development circadian sleep/wake cycle eating behavior response to peptide hormone response to leptin response to ethanol response to ether response to pyrethroid response to steroid hormone embryonic camera-type eye morphogenesis cognition response to corticosterone response to electrical stimulus response to growth factor cellular response to manganese ion cellular response to alkaloid cellular response to nicotine cellular response to glucose stimulus cellular response to growth factor stimulus cellular response to xenobiotic stimulus response to curcumin hyaloid vascular plexus regression
          It has to be done as per old AB suggested Products section.

          Disclaimer

          Close

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-5db85fbf84-lw26s:80/100.66.78.224:80.
          git-commit: 64d6f4084a597c0a860dbc264f43a6f389f0d7ac
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.60.1-2026-secondary-format-patch